Gene Transcripts

Gene Symbol
Rrp1
Entrez Gene ID
309674
Gene Name
ribosomal RNA processing 1
Gene Aliases
Nnp1
Chromosome Location
Chr. 20: 10982016 - 10993260 on Build Rnor_6.0
UniGene ID
Rn.104084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012073.1 NP_001012073.1 7 634
GenBank mRNA BC087152.1 - 7 634

View Full Product Details