Gene Transcripts

Gene Symbol
Defa10
Entrez Gene ID
613221
Gene Name
defensin alpha 10
Gene Aliases
Chromosome Location
Chr. 16: 75498213 - 75500336 on Build Rnor_6.0
UniGene ID
Rn.215840
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033074.1 NP_001028246.1 2 225
XM_008771364.1 XP_008769586.1 3 426
XM_008771365.1 XP_008769587.1 3 423
GenBank mRNA AY623754.1 - 2 225

View Full Product Details