Gene Transcripts

Gene Symbol
Acvr2b
Entrez Gene ID
25366
Gene Name
activin A receptor, type IIB
Gene Aliases
Chromosome Location
Chr. 8: 128087315 - 128118844 on Build Rnor_6.0
UniGene ID
Rn.24240
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031554.1 NP_113742.1 3 538
XM_006244072.2 XP_006244134.1 3 568
GenBank mRNA BC097358.1 - 3 538
M87067.1 - 3 775

View Full Product Details