Gene Transcripts

Gene Symbol
Hbq1b
Entrez Gene ID
544763
Gene Name
hemoglobin, theta 1B
Gene Aliases
F830116E18Rik, Hbqt1
Chromosome Location
Chr. 11: 32286702 - 32296033 on Build GRCm38
UniGene ID
Mm.450958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033981.3 NP_001029153.1 3 499
GenBank mRNA AK157060.1 - 3 500
BC141224.1 - 3 477 9055836

View Full Product Details