Gene Transcripts

Gene Symbol
Samd4
Entrez Gene ID
74480
Gene Name
sterile alpha motif domain containing 4
Gene Aliases
1700024G08Rik, 1700111L17Rik, 4933436G17Rik, Samd4a, Smaug, Smaug1
Chromosome Location
Chr. 14: 46882965 - 47105817 on Build GRCm38
UniGene ID
Mm.214454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037221.2 NP_001032298.1 13 2915
NM_001163433.1 NP_001156905.1 12 2395
NM_028966.3 NP_083242.1 12 2651
GenBank mRNA AK017081.1 - 8 1229
AK034323.1 - 11 2245
AK133470.1 - 12 2631
AK134243.1 - 13 2913
AK161277.1 - 12 2389
BC049740.1 - 8 1201
BC145827.1 - 12 2517 40131130
BC145829.1 - 12 2516 40131132
DQ278487.1 - 12 2428

View Full Product Details