Gene Transcripts

Gene Symbol
Snrnp70
Entrez Gene ID
20637
Gene Name
small nuclear ribonucleoprotein 70 (U1)
Gene Aliases
2700022N21Rik, 3200002N22Rik, AI325098, R74807, Rnu1p70, Rnulp70, Snrp70, Srnp70, U1-70, U1-70K
Chromosome Location
Chr. 7: 45376454 - 45395742 on Build GRCm38
UniGene ID
Mm.216386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005045.2 - 6, 7 584
AK011564.1 - 6, 7 540
AK012272.1 - 4, 5 233
AK014311.2 - 6, 7 581
AK077425.1 - 6, 7 586
AK133115.1 - 6, 7 585
AK146729.1 - 6, 7 585
AK148013.1 - 6, 7 611
BC049128.1 - 6, 7 550
BC051414.1 - 5, 6 385
BC131994.1 - 6, 7 477 40130271
BC132020.1 - 6, 7 477 40130297

View Full Product Details