Gene Transcripts

Gene Symbol
Tubb2a
Entrez Gene ID
22151
Gene Name
tubulin, beta 2A class IIA
Gene Aliases
M(beta)2, Tubb2
Chromosome Location
Chr. 13: 34074278 - 34078008 on Build GRCm38
UniGene ID
Mm.422827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009450.2 NP_033476.1 4 683
GenBank mRNA AK134545.1 - 4 682
BC003475.1 - 1 163
BC055441.1 - 4 673 4016867

View Full Product Details