Gene Transcripts

Gene Symbol
Klra9
Entrez Gene ID
16640
Gene Name
killer cell lectin-like receptor subfamily A, member 9
Gene Aliases
LY49I1, Ly49I
Chromosome Location
Chr. 6: 130178626 - 130194374 on Build GRCm38
UniGene ID
Mm.457996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010651.3 NP_034781.2 3 292
GenBank mRNA AF237686.1 - 216
AY860970.1 - 2 129
FJ159530.1 - 129
U34891.1 - 2 129
U34892.1 - 129
U49865.1 - 292
U49866.1 - 292
U49867.1 - 292

View Full Product Details