Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
58234
Gene Name
SH3/ankyrin domain gene 3
Gene Aliases
AI841104, SHANK3c-3, SHANK3c-4, Shank3b
Chromosome Location
Chr. 15: 89499707 - 89560261 on Build GRCm38
UniGene ID
Mm.146855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021423.3 NP_067398.2 21 4479
GenBank mRNA AB231013.1 - 21 4479
AB841411.1 - 11 3443
AK173228.1 - 1 1858
KF612595.1 - 1 27

View Full Product Details