Gene Transcripts

Gene Symbol
Clec4g
Entrez Gene ID
75863
Gene Name
C-type lectin domain family 4, member g
Gene Aliases
4930572L20Rik
Chromosome Location
Chr. 8: 3707069 - 3721183 on Build GRCm38
UniGene ID
Mm.109183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029465.3 NP_083741.1 5 312
GenBank mRNA AK079992.1 - 5 313
BC025069.1 - 5 304
BC087963.1 - 5 312 6766377

View Full Product Details