Gene Transcripts
- Gene Symbol
- KIAA1524
- Entrez Gene ID
- 57650
- Gene Name
- KIAA1524
- Gene Aliases
- CIP2A, p90
- Chromosome Location
- Chr. 3: 108545752 - 108589644 on Build GRCh38
- UniGene ID
- Hs.591308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020890.2 |
NP_065941.2
|
1, 2 |
379 |
|
XM_006713716.3 |
XP_006713779.1
|
1, 2 |
166 |
|
XM_011513056.1 |
XP_011511358.1
|
1, 2 |
187 |
|
GenBank mRNA |
AF334474.1 |
- |
1, 2 |
178 |
|
AK056933.1 |
- |
1, 2 |
188 |
|
AK308315.1 |
- |
1, 2 |
189 |
|
AK310446.1 |
- |
1, 2 |
383 |
|
AL713689.1 |
- |
1, 2 |
174 |
|
BC063433.1 |
- |
1, 2 |
170 |
|
BC082961.1 |
- |
1, 2 |
132 |
|
BC130564.1 |
- |
1, 2 |
146 |
40146595 |
BC136371.1 |
- |
1, 2 |
146 |
9020358 |
View Full Product Details