Gene Transcripts

Gene Symbol
Wdr18
Entrez Gene ID
216156
Gene Name
WD repeat domain 18
Gene Aliases
2310012I10Rik, AU044733, AW122032
Chromosome Location
Chr. 10: 79960152 - 79969246 on Build GRCm38
UniGene ID
Mm.35828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175450.4 NP_780659.2 10 2928
GenBank mRNA AK082695.1 - 10 2931
AK133716.1 - 10 2929
AK166113.1 - 2929
AK168691.1 - 10 2926

View Full Product Details