Gene Transcripts

Gene Symbol
Tceal5
Entrez Gene ID
331532
Gene Name
transcription elongation factor A (SII)-like 5
Gene Aliases
6430401A05Rik
Chromosome Location
Chr. X: 136200948 - 136203851 on Build GRCm38
UniGene ID
Mm.41605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177919.2 NP_808587.1 3 866
GenBank mRNA AK032140.1 - 3 868
BC100415.1 - 3 891 6535676
BC115460.1 - 2 777 40100235
BC115461.1 - 2 777 40100237

View Full Product Details