Gene Transcripts
- Gene Symbol
- DPP10
- Entrez Gene ID
- 57628
- Gene Name
- dipeptidyl peptidase like 10
- Gene Aliases
- DPL2, DPPY, DPRP-3, DPRP3
- Chromosome Location
- Chr. 2: 114442358 - 115845752 on Build GRCh38
- UniGene ID
- Hs.580539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001004360.3 |
NP_001004360.2
|
3, 4 |
369 |
|
NM_001178034.1 |
NP_001171505.1
|
3, 4 |
296 |
|
NM_001178036.1 |
NP_001171507.1
|
4, 5 |
773 |
|
NM_001178037.1 |
NP_001171508.1
|
3, 4 |
339 |
|
NM_001321905.1 |
NP_001308834.1
|
4, 5 |
799 |
|
NM_001321906.1 |
NP_001308835.1
|
4, 5 |
415 |
|
NM_001321907.1 |
NP_001308836.1
|
3, 4 |
684 |
|
NM_001321908.1 |
NP_001308837.1
|
3, 4 |
658 |
|
NM_001321910.1 |
NP_001308839.1
|
4, 5 |
742 |
|
NM_001321911.1 |
NP_001308840.1
|
3, 4 |
741 |
|
NM_001321912.1 |
NP_001308841.1
|
4, 5 |
773 |
|
NM_001321913.1 |
NP_001308842.1
|
2, 3 |
569 |
|
NM_001321914.1 |
NP_001308843.1
|
3, 4 |
299 |
|
NM_020868.4 |
NP_065919.2
|
3, 4 |
684 |
|
XM_017004565.1 |
XP_016860054.1
|
4, 5 |
843 |
|
XM_017004566.1 |
XP_016860055.1
|
2, 3 |
288 |
|
GenBank mRNA |
AB040925.2 |
- |
3, 4 |
251 |
|
AK055256.1 |
- |
5, 6 |
857 |
|
AK289967.1 |
- |
3, 4 |
383 |
|
AK296490.1 |
- |
3, 4 |
371 |
|
AK315147.1 |
- |
3, 4 |
488 |
|
AY172661.3 |
- |
3, 4 |
263 |
|
AY387785.1 |
- |
3, 4 |
285 |
|
BC030832.1 |
- |
3, 4 |
720 |
4811771 |
BI597037.1 |
- |
3, 4 |
326 |
|
DQ857321.1 |
- |
3, 4 |
251 |
|
DQ857322.1 |
- |
3, 4 |
275 |
|
DQ857323.1 |
- |
3, 4 |
242 |
|
View Full Product Details