Gene Transcripts

Gene Symbol
PLEKHA4
Entrez Gene ID
57664
Gene Name
pleckstrin homology domain containing A4
Gene Aliases
PEPP1
Chromosome Location
Chr. 19: 48837092 - 48868627 on Build GRCh38
UniGene ID
Hs.9469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161354.1 NP_001154826.1 10 1633
NM_020904.2 NP_065955.2 11 1708
XM_005259107.2 XP_005259164.1 10 1628
XM_006723301.2 XP_006723364.1 10 1628
XM_011527157.1 XP_011525459.1 11 1715
XM_011527158.2 XP_011525460.1 10 1554
XM_011527159.1 XP_011525461.1 11 1728
XM_011527160.1 XP_011525462.1 11 1715
XM_011527162.2 XP_011525464.1 4 324
XM_017027037.1 XP_016882526.1 8 992
GenBank mRNA AB208908.1 - 11 1670
AY007233.1 - 11 1682
BC024157.2 - 11 1688 3953478
BC033832.1 - 3 158
BC064601.1 - 10 1460 5785991

View Full Product Details