Gene Transcripts

Gene Symbol
HIST1H2AG
Entrez Gene ID
8969
Gene Name
histone cluster 1, H2ag
Gene Aliases
H2A.1b, H2A/p, H2AFP, H2AG, pH2A/f
Chromosome Location
Chr. 6: 27133038 - 27133535 on Build GRCh38
UniGene ID
Hs.51011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021064.4 NP_066408.1 1 426
GenBank mRNA BC016677.1 - 1 411
L19778.1 - 1 422

View Full Product Details