Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001258461.1 | NP_001245390.1 | 5 | 926 | |
NM_006712.4 | NP_006703.1 | 5 | 1007 | ||
NM_033015.3 | NP_148936.2 | 4 | 584 | ||
XM_005249932.1 | XP_005249989.1 | 5 | 1199 | ||
XM_005249933.3 | XP_005249990.1 | 5 | 1405 | ||
XM_011515761.2 | XP_011514063.1 | 5 | 1007 | ||
XM_011515762.2 | XP_011514064.1 | 5 | 1007 | ||
XM_011515763.1 | XP_011514065.1 | 4 | 465 | ||
XM_017011704.1 | XP_016867193.1 | 5 | 1118 | ||
XM_017011705.1 | XP_016867194.1 | 4 | 562 | ||
XM_017011706.1 | XP_016867195.1 | 4 | 754 | ||
GenBank mRNA | AB209783.1 | - | 5 | 827 | |
AK023141.1 | - | 3 | 1073 | ||
AK223132.1 | - | 5 | 917 | ||
AK292232.1 | - | 5 | 892 | ||
AL713798.1 | - | 3 | 1860 | ||
BC000377.2 | - | 5 | 919 | 2820338 | |
BC006386.2 | - | 4 | 890 | ||
BC011770.2 | - | 5 | 918 | 3831196 | |
BC039026.2 | - | 4 | 554 | 4940575 | |
BM799420.1 | - | 2 | 255 | ||
X86779.1 | - | 5 | 911 |