Gene Transcripts
- Gene Symbol
- DHRS4L1
- Entrez Gene ID
- 728635
- Gene Name
- dehydrogenase/reductase 4 like 1
- Gene Aliases
- SDR25C4
- Chromosome Location
- Chr. 14: 24007009 - 24051377 on Build GRCh38
- UniGene ID
- Hs.647569
- Species
- Homo sapiens
- Gene Symbol
- DHRS4
- Entrez Gene ID
- 10901
- Gene Name
- dehydrogenase/reductase 4
- Gene Aliases
- CR, NRDR, PHCR, PSCD, SCAD-SRL, SDR-SRL, SDR25C1, SDR25C2
- Chromosome Location
- Chr. 14: 23953735 - 23969279 on Build GRCh38
- UniGene ID
- Hs.528385
- Species
- Homo sapiens
- Gene Symbol
- DHRS4L2
- Entrez Gene ID
- 317749
- Gene Name
- dehydrogenase/reductase 4 like 2
- Gene Aliases
- SDR25C3
- Chromosome Location
- Chr. 14: 23969874 - 24006408 on Build GRCh38
- UniGene ID
- Hs.743442
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021587.1 |
- |
1 |
744 |
|
GU564234.1 |
- |
7 |
621 |
|
GU564235.1 |
- |
6 |
723 |
|
View Full Product Details