Gene Transcripts
- Gene Symbol
- ADCY1
- Entrez Gene ID
- 107
- Gene Name
- adenylate cyclase 1
- Gene Aliases
- AC1, DFNB44
- Chromosome Location
- Chr. 7: 45574140 - 45723116 on Build GRCh38
- UniGene ID
- Hs.192215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021116.2 |
NP_066939.1
|
20 |
11230 |
|
GenBank mRNA |
AK055343.1 |
- |
1 |
1635 |
|
View Full Product Details