Gene Transcripts

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
Chromosome Location
Chr. 19: 40393762 - 40414718 on Build GRCh38
UniGene ID
Hs.205457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020956.2 NP_066007.1 6 3800
NM_181882.2 NP_870998.2 7 3151
XM_011527171.2 XP_011525473.1 7 3143
XM_017027046.1 XP_016882535.1 3 3302
XM_017027047.1 XP_016882536.1 4 2845
GenBank mRNA AB046840.1 - 3 3281
AF321191.1 - 7 3151
AF321192.1 - 6 3800
BC067266.1 - 7 3153 6177899

View Full Product Details