Gene Transcripts

Gene Symbol
METTL14
Entrez Gene ID
57721
Gene Name
methyltransferase like 14
Gene Aliases
Chromosome Location
Chr. 4: 118685370 - 118712270 on Build GRCh38
UniGene ID
Hs.596872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020961.3 NP_066012.1 3, 4 416
XM_005263166.4 XP_005263223.1 3, 4 411
XM_011532162.2 XP_011530464.1 3, 4 411
GenBank mRNA AB046847.1 - 3, 4 371
AK055555.1 - 3, 4 394
AK296056.1 - 1, 2 36
BC006565.2 - 3, 4 367 3010654
BC007449.2 - 3, 4 367 3010654
BG773151.1 - 3, 4 421

View Full Product Details