Gene Transcripts

Gene Symbol
RTN4RL2
Entrez Gene ID
349667
Gene Name
reticulon 4 receptor-like 2
Gene Aliases
NGRH1, NgR2
Chromosome Location
Chr. 11: 57460537 - 57477539 on Build GRCh38
UniGene ID
Hs.502618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178570.2 NP_848665.1 2 478
GenBank mRNA AF532858.1 - 2 149
AX751514.1 - 2 149
AY250221.1 - 2 149
BC113673.1 - 2 171 8322725
BC117276.1 - 2 171 40125827
BC143818.1 - 2 171 9052333
BU845536.1 - 478
DQ864979.1 - 2 149

View Full Product Details