Gene Transcripts

Gene Symbol
KRT40
Entrez Gene ID
125115
Gene Name
keratin 40
Gene Aliases
CK-40, K40, KA36
Chromosome Location
Chr. 17: 40977716 - 40987135 on Build GRCh38
UniGene ID
Hs.534929
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182497.3 NP_872303.2 3 580
XM_011524339.2 XP_011522641.2 1 2347
XM_017024189.1 XP_016879678.1 1 2347
GenBank mRNA AJ786658.1 - 1 420
AK093919.1 - 3 581
BC130396.1 - 2 455 40146427
BC144250.1 - 2 455 9052770
BC150174.1 - 3 580 40147687
BC150191.1 - 3 580 40148622

View Full Product Details