Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
10218
Gene Name
angiopoietin like 7
Gene Aliases
AngX, CDT6, dJ647M16.1
Chromosome Location
Chr. 1: 11189289 - 11195981 on Build GRCh38
UniGene ID
Hs.146559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021146.3 NP_066969.1 1 449
XM_017000004.1 XP_016855493.1 1 414
GenBank mRNA AB593026.1 - 1 383
AB593027.1 - 1 450
AK313716.1 - 1 415
AY358301.1 - 1 372
BC001881.1 - 1 397 3544149
BT009802.1 - 1 158
CA391009.1 - 1 449
Y16132.1 - 1 165

View Full Product Details