Gene Transcripts

Gene Symbol
CCAR2
Entrez Gene ID
57805
Gene Name
cell cycle and apoptosis regulator 2
Gene Aliases
DBC-1, DBC1, KIAA1967, NET35, p30 DBC, p30DBC
Chromosome Location
Chr. 8: 22604632 - 22620471 on Build GRCh38
UniGene ID
Hs.744848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021174.5 NP_066997.3 4 664
NR_033902.1 4 482
XM_011544603.1 XP_011542905.1 4 482
XM_011544604.1 XP_011542906.1 4 482
XM_017013717.1 XP_016869206.1 4 664
GenBank mRNA AB075847.1 - 1 13
AK095712.1 - 4 338
AK314535.1 - 4 357
AL834162.1 - 4 651
AL834351.1 - 4 652
AL834352.1 - 4 652
BC065495.1 - 4 340 5496068
BI561261.1 - 4 379
BX640952.1 - 4 351

View Full Product Details