Gene Transcripts

Gene Symbol
RNPEPL1
Entrez Gene ID
57140
Gene Name
arginyl aminopeptidase like 1
Gene Aliases
Chromosome Location
Chr. 2: 240568526 - 240578732 on Build GRCh38
UniGene ID
Hs.5345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018226.4 NP_060696.4 11 2988
XM_005247036.4 XP_005247093.1 11 3016
GenBank mRNA AF300795.1 - 11 2694
AK001668.1 - 7 1952
AK130185.1 - 4 1556
AK130805.1 - 4 3922
AL512754.1 - 4 1573
BC017301.2 - 8 2140
BC067258.1 - 11 2563 6183046
BC082975.1 - 11 2888 6379537
BC099906.1 - 9 3101 5311883

View Full Product Details