Gene Transcripts

Gene Symbol
PLEKHB1
Entrez Gene ID
58473
Gene Name
pleckstrin homology domain containing B1
Gene Aliases
KPL1, PHR1, PHRET1
Chromosome Location
Chr. 11: 73646178 - 73662819 on Build GRCh38
UniGene ID
Hs.445489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130033.1 NP_001123505.1 7 1255
NM_001130034.1 NP_001123506.1 8 1101
NM_001130035.1 NP_001123507.1 7 996
NM_001130036.1 NP_001123508.1 6 1128
NM_021200.2 NP_067023.1 8 1360
XM_011545192.2 XP_011543494.1 8 1934
XM_011545193.2 XP_011543495.1 7 1829
GenBank mRNA AB209716.1 - 7 908
AF081583.1 - 7 877
AF093249.1 - 7 880
AF100612.1 - 8 985
AF101054.1 - 7 1251
AK054725.1 - 3 1013
AK289620.1 - 7 908
AK289909.1 - 7 874
AK296033.1 - 7 800
AK311158.1 - 8 753
BC008075.2 - 7 934 3050369
EB387039.1 - 1 511
U89715.1 - 8 1356

View Full Product Details