Gene Transcripts

Gene Symbol
RHOU
Entrez Gene ID
58480
Gene Name
ras homolog family member U
Gene Aliases
ARHU, CDC42L1, G28K, WRCH1, hG28K
Chromosome Location
Chr. 1: 228644647 - 228746669 on Build GRCh38
UniGene ID
Hs.647774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021205.5 NP_067028.1 3 1313
NR_037962.1 3 771
GenBank mRNA AB051826.1 - 3 726
AB074878.1 - 3 786
AB209511.1 - 4 845
AF211836.1 - 3 1269
AF251701.1 - 3 506
AF282258.1 - 3 886
AF378087.1 - 3 647
AK289971.1 - 3 899
BC040076.1 - 3 894 5768957

View Full Product Details