Gene Transcripts

Gene Symbol
NTN4
Entrez Gene ID
59277
Gene Name
netrin 4
Gene Aliases
PRO3091
Chromosome Location
Chr. 12: 95657807 - 95790758 on Build GRCh38
UniGene ID
Hs.201034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021229.3 NP_067052.2 8 1984
GenBank mRNA AF119916.3 - 9 1521
AF278532.1 - 8 1986
AF297711.1 - 8 1836
AK024691.1 - 7 1060
AK026833.1 - 3 330
AK226070.1 - 8 1841
AK290591.1 - 8 1521
AK315796.1 - 8 1843
AL137540.1 - 6 852
AL833767.1 - 7 1388
BC013591.1 - 7 1059
BC136798.1 - 8 1959 9020788
BC136799.1 - 8 1959 9020789

View Full Product Details