Gene Transcripts

Gene Symbol
PIEZO2
Entrez Gene ID
63895
Gene Name
piezo type mechanosensitive ion channel component 2
Gene Aliases
C18orf30, C18orf58, DA3, DA5, FAM38B, FAM38B2, HsT748, HsT771, MWKS
Chromosome Location
Chr. 18: 10670237 - 11149561 on Build GRCh38
UniGene ID
Hs.436902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022068.3 NP_071351.2 43 6959
XM_011525723.2 XP_011524025.1 46 7889
XM_011525724.2 XP_011524026.1 45 7832
XM_011525725.2 XP_011524027.1 44 7799
XM_011525726.2 XP_011524028.1 45 7706
XM_017025918.1 XP_016881407.1 45 7850
GenBank mRNA AK027056.1 - 3 366
AK302764.1 - 5 796
JN790819.1 - 43 6793

View Full Product Details