Gene Transcripts

Gene Symbol
GLIPR2
Entrez Gene ID
152007
Gene Name
GLI pathogenesis related 2
Gene Aliases
C9orf19, GAPR-1, GAPR1
Chromosome Location
Chr. 9: 36136536 - 36163913 on Build GRCh38
UniGene ID
Hs.493819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287010.1 NP_001273939.1 2, 3 365
NM_001287011.1 NP_001273940.1 2, 3 365
NM_001287013.1 NP_001273942.1 2, 3 202
NM_022343.3 NP_071738.1 2, 3 365
NR_104637.1 4, 5 746
NR_104638.1 2, 3 157
NR_104639.1 2, 3 365
GenBank mRNA AJ011129.1 - 1, 2 122
AK074488.1 - 2, 3 187
AK075091.1 - 2, 3 166
AK091288.1 - 3, 4 979
AK298741.1 - 2, 3 166
AK307243.1 - 2, 3 166
AY039756.1 - 2, 3 156
BC017918.1 - 2, 3 168 4296721
BF515272.1 - 2, 3 365
CR749781.1 - 4, 5 547
DA268281.1 - 2, 3 202
DA271525.1 - 2, 3 142
DA986727.1 - 2, 3 166

View Full Product Details