Gene Transcripts

Gene Symbol
SPATA1
Entrez Gene ID
100505741
Gene Name
spermatogenesis associated 1
Gene Aliases
SP-2, SPAP1
Chromosome Location
Chr. 1: 84506301 - 84556495 on Build GRCh38
UniGene ID
Hs.584956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310156.1 NP_001297085.1 3 333
XM_011542514.2 XP_011540816.1 3 333
XM_011542515.2 XP_011540817.1 3 333
XM_011542516.1 XP_011540818.1 3 333
XM_011542517.1 XP_011540819.1 3 333
XM_011542519.2 XP_011540821.1 3 333
XM_011542520.2 XP_011540822.1 3 333
XM_011542521.2 XP_011540823.1 3 333
XM_011542522.2 XP_011540824.1 3 333
GenBank mRNA AF306347.1 - 3 305
BC119641.1 - 3 237
BC119642.1 - 3 237
BC127634.1 - 3 237 40115717
CD300599.1 - 3 349

View Full Product Details