Gene Transcripts

Gene Symbol
STRA6
Entrez Gene ID
64220
Gene Name
stimulated by retinoic acid 6
Gene Aliases
MCOPCB8, MCOPS9, PP14296
Chromosome Location
Chr. 15: 74179466 - 74212267 on Build GRCh38
UniGene ID
Hs.24553
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142617.1 NP_001136089.1 9 998
NM_001142618.1 NP_001136090.1 9 980
NM_001142619.1 NP_001136091.1 9 971
NM_001199040.1 NP_001185969.1 9 1114
NM_001199041.1 NP_001185970.1 9 1125
NM_001199042.1 NP_001185971.1 9 1229
NM_022369.3 NP_071764.3 9 934
XM_011521883.1 XP_011520185.1 9 900
XM_011521884.1 XP_011520186.1 9 1041
XM_011521885.2 XP_011520187.1 9 875
XM_017022478.1 XP_016877967.1 9 849
XM_017022479.1 XP_016877968.1 9 940
XM_017022480.1 XP_016877969.1 9 964
GenBank mRNA AF352728.1 - 9 800
AF352729.1 - 9 910
AF370419.2 - 9 906
AK022603.1 - 5 484
AK056125.1 - 9 995
AK091152.1 - 9 1024
AK092227.1 - 4 461
AK291966.1 - 9 870
AK298921.1 - 9 869
AK298928.1 - 7 538
AK299191.1 - 9 1114
AK302932.1 - 9 1125
AK309676.1 - 9 1209
AK310878.1 - 9 857
AY358748.1 - 9 910
AY359089.1 - 9 800
BC025256.1 - 9 906 4892821
BX537413.1 - 9 876

View Full Product Details