Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040284.2 | NP_001035374.2 | 13 | 2060 | |
NM_001040285.2 | NP_001035375.2 | 13 | 1919 | ||
XM_005256092.4 | XP_005256149.1 | 12 | 2243 | ||
XM_011523275.2 | XP_011521577.1 | 13 | 2056 | ||
XM_011523276.2 | XP_011521578.1 | 13 | 1917 | ||
XM_011523278.2 | XP_011521580.1 | 12 | 1531 | ||
XM_017023555.1 | XP_016879044.1 | 13 | 2116 | ||
GenBank mRNA | AF089897.1 | - | 9 | 1172 | |
AK001141.1 | - | 9 | 1139 | ||
AK223410.1 | - | 11 | 1330 | ||
AK299294.1 | - | 6 | 872 | ||
AK300918.1 | - | 13 | 2060 | ||
AL833922.1 | - | 10 | 1162 | ||
BC089405.1 | - | 8 | 1103 | ||
FR872509.1 | - | 12 | 1824 |