Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr. 17: 50547089 - 50555852 on Build GRCh38
UniGene ID
Hs.103147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258372.1 NP_001245301.1 2 273
NM_001258373.1 NP_001245302.1 3 376
NM_022827.3 NP_073738.2 3 321
GenBank mRNA AK025000.1 - 2 170
AK092731.1 - 3 306
AK125807.1 - 5 1051
AK225189.1 - 2 170
AK291805.1 - 3 264
AK302850.1 - 2 220
AK308616.1 - 3 446
AK310416.1 - 2 165
BC025255.1 - 3 209 5019010
BC065526.1 - 3 263 6149633
BC111029.1 - 2 178 5245538
BM787432.1 - 2 236
CR936769.1 - 2 191
DA295930.1 - 3 336
DA489234.1 - 3 321
DQ238597.1 - 2 239
GQ472220.1 - 2 178

View Full Product Details