Gene Transcripts

Gene Symbol
UPF3A
Entrez Gene ID
65110
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Gene Aliases
HUPF3A, RENT3A, UPF3
Chromosome Location
Chr. 13: 114281584 - 114305817 on Build GRCh38
UniGene ID
Hs.533855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023011.3 NP_075387.1 7 783
NM_080687.2 NP_542418.1 6 684
XM_005266202.4 XP_005266259.1 4 409
XM_006719991.3 XP_006720054.1 6 658
XM_011534844.1 XP_011533146.1 5 551
XM_011534845.2 XP_011533147.1 7 1311
XM_011534846.1 XP_011533148.1 7 730
XM_011534847.2 XP_011533149.1 6 1123
XM_011534848.2 XP_011533150.1 7 1268
XM_017020709.1 XP_016876198.1 6 1495
XM_017020710.1 XP_016876199.1 7 880
XM_017020711.1 XP_016876200.1 6 781
GenBank mRNA AF318575.1 - 7 676
AK001069.1 - 2 60
AK097027.1 - 3 745
AK302003.1 - 7 719
AY013250.1 - 7 783
BC008694.1 - 3 196
BC023569.1 - 6 648 4127465

View Full Product Details