Gene Transcripts

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
Chromosome Location
Chr. 7: 135092303 - 135170826 on Build GRCh38
UniGene ID
Hs.643113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243749.1 NP_001230678.1 3 785
NM_001243751.1 NP_001230680.1 3 532
NM_001243752.1 NP_001230681.1 3 606
NM_001243753.1 NP_001230682.1 3 410
NM_001243754.1 NP_001230683.1 3 312
NM_001243755.1 NP_001230684.1 3 228
NM_024033.3 NP_076938.2 3 495
XM_017012589.1 XP_016868078.1 3 618
XM_017012590.1 XP_016868079.1 3 544
XM_017012591.1 XP_016868080.1 3 398
XM_017012593.1 XP_016868082.1 3 448
XM_017012594.1 XP_016868083.1 3 655
XM_017012595.1 XP_016868084.1 3 527
GenBank mRNA AK026103.1 - 3 621
AK130795.1 - 1 321
AL708733.1 - 3 228
BC000168.2 - 3 364 2900206
BC067350.1 - 3 401 6568468
BG770658.1 - 3 395
BM727569.1 - 2 326
BP213903.1 - 3 393
BQ423284.1 - 245
CB961457.1 - 3 522
DA213716.1 - 3 489
DB129229.1 - 3 486

View Full Product Details