Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023079.4 | NP_075567.2 | 7 | 1739 | |
GenBank mRNA | AK023917.1 | - | 6 | 1277 | |
AK024030.1 | - | 8 | 1594 | ||
AK091644.1 | - | 5 | 2870 | ||
AK122770.1 | - | 7 | 1637 | ||
AK130303.1 | - | 8 | 1597 | ||
AL713782.1 | - | 3 | 894 | ||
BC015169.1 | - | 6 | 1266 | ||
BC015890.1 | - | 7 | 1607 | 3915698 |