Gene Transcripts

Gene Symbol
CYP4F12
Entrez Gene ID
66002
Gene Name
cytochrome P450 family 4 subfamily F member 12
Gene Aliases
CYPIVF12, F22329_1
Chromosome Location
Chr. 19: 15673018 - 15698819 on Build GRCh38
UniGene ID
Hs.131459
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023944.3 NP_076433.3 8 1060
NR_117085.1 7 1078
XM_006722850.3 XP_006722913.1 8 945
XM_011528202.2 XP_011526504.1 6 580
XM_011528203.2 XP_011526505.1 5 1802
XM_011528204.2 XP_011526506.1 6 1281
XM_011528205.2 XP_011526507.1 5 493
XM_011528207.2 XP_011526509.1 5 2189
XM_011528208.2 XP_011526510.1 7 699
XM_017027172.1 XP_016882661.1 8 1002
GenBank mRNA AB035130.1 - 8 971
AB035131.1 - 8 971
AK074412.1 - 2 2362
AK075435.1 - 7 1006
AL832171.1 - 7 1967
AY008841.1 - 7 941
AY358977.1 - 8 982
BC035350.1 - 8 987 5187264

View Full Product Details