Gene Transcripts

Gene Symbol
NOTCH2
Entrez Gene ID
4853
Gene Name
notch 2
Gene Aliases
AGS2, HJCYS, hN2
Chromosome Location
Chr. 1: 119911553 - 120073449 on Build GRCh38
UniGene ID
Hs.487360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024408.3 NP_077719.2 34 8337
XM_005270901.3 XP_005270958.1 35 10651
XM_011541519.2 XP_011539821.1 34 13445
XM_011541520.2 XP_011539822.1 34 10355
XM_017001372.1 XP_016856861.1 34 8007
XM_017001373.1 XP_016856862.1 34 7986
GenBank mRNA AF308601.1 - 34 8052
AF315356.1 - 34 8052
AK027869.1 - 8 3175
AL133036.1 - 1 2014

View Full Product Details