Gene Transcripts

Gene Symbol
PRDM14
Entrez Gene ID
63978
Gene Name
PR domain 14
Gene Aliases
PFM11
Chromosome Location
Chr. 8: 70051126 - 70071327 on Build GRCh38
UniGene ID
Hs.736037
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024504.3 NP_078780.1 2 704
GenBank mRNA AF319458.1 - 2 704
AK022595.1 - 2 704
BC052311.1 - 2 617 6671984
KJ463837.1 - 1 502

View Full Product Details