Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr. 14: 100376410 - 100530303 on Build GRCh38
UniGene ID
Hs.497600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161476.1 NP_001154948.1 4 1158
NM_024515.4 NP_078791.3 4 1289
XM_005268056.4 XP_005268113.1 4 1165
XM_005268057.3 XP_005268114.1 3 315
XM_005268058.4 XP_005268115.1 3 329
XM_011537150.2 XP_011535452.1 4 1412
XM_011537151.2 XP_011535453.2 3 768
XM_017021639.1 XP_016877128.1 3 317
GenBank mRNA AK291960.1 - 4 1121
AY295082.1 - 4 1172
BC003641.2 - 3 395 3529568
BC007953.2 - 2 942
CT003969.1 - 3 321

View Full Product Details