Gene Transcripts

Gene Symbol
C22orf46
Entrez Gene ID
79640
Gene Name
chromosome 22 open reading frame 46
Gene Aliases
Chromosome Location
Chr. 22: 41688939 - 41698136 on Build GRCh38
UniGene ID
Hs.517653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142964.1 NP_001136436.1 2 4915
XM_011530373.2 XP_011528675.1 3 4823
XM_017028939.1 XP_016884428.1 4 4954
GenBank mRNA AK027237.1 - 1 1828
AK095008.1 - 1 3670
AK131457.1 - 4 4749
BC007210.1 - 1 1910 3344580
BC035403.1 - 2 4916

View Full Product Details