Gene Transcripts

Gene Symbol
MANEA
Entrez Gene ID
79694
Gene Name
mannosidase endo-alpha
Gene Aliases
ENDO, hEndo
Chromosome Location
Chr. 6: 95577497 - 95609452 on Build GRCh38
UniGene ID
Hs.734632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024641.3 NP_078917.2 5 946
XM_005267147.3 XP_005267204.1 5 1339
GenBank mRNA AJ577574.1 - 4 811
AK022900.1 - 4 452
AK291940.1 - 5 877
AY372528.1 - 4 766
BC137014.1 - 4 815 9021007
BC137016.1 - 4 815 9021009
BC146671.1 - 5 881 40148201
BC150199.1 - 5 881 40148811
BX537398.1 - 5 877
BX640869.1 - 5 907
CR936827.1 - 5 881

View Full Product Details