Gene Transcripts
- Gene Symbol
- TBL1XR1
- Entrez Gene ID
- 79718
- Gene Name
- transducin (beta)-like 1 X-linked receptor 1
- Gene Aliases
- C21, DC42, IRA1, MRD41, TBLR1
- Chromosome Location
- Chr. 3: 177019355 - 177197357 on Build GRCh38
- UniGene ID
- Hs.714201
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321193.1 |
NP_001308122.1
|
4, 5 |
478 |
|
NM_001321194.1 |
NP_001308123.1
|
5, 6 |
658 |
|
NM_001321195.1 |
NP_001308124.1
|
3, 4 |
375 |
|
NM_024665.5 |
NP_078941.2
|
4, 5 |
539 |
|
XM_005247775.2 |
XP_005247832.1
|
3, 4 |
494 |
|
XM_006713746.1 |
XP_006713809.1
|
5, 6 |
548 |
|
XM_011513141.1 |
XP_011511443.1
|
5, 6 |
482 |
|
XM_011513142.2 |
XP_011511444.1
|
4, 5 |
463 |
|
XM_011513143.2 |
XP_011511445.1
|
3, 4 |
389 |
|
XM_017007185.1 |
XP_016862674.1
|
4, 5 |
476 |
|
GenBank mRNA |
AF268193.1 |
- |
4, 5 |
372 |
|
AF314544.1 |
- |
3, 4 |
338 |
|
AK022956.1 |
- |
4, 5 |
438 |
|
AK296946.1 |
- |
3, 4 |
273 |
|
BC060320.1 |
- |
3, 4 |
392 |
|
BC113421.1 |
- |
3, 4 |
281 |
8322473 |
BX648935.1 |
- |
4, 5 |
473 |
|
DA159105.1 |
- |
3, 4 |
324 |
|
DA460721.1 |
- |
2, 3 |
300 |
|
View Full Product Details