Gene Transcripts

Gene Symbol
NIPAL2
Entrez Gene ID
79815
Gene Name
NIPA like domain containing 2
Gene Aliases
NPAL2
Chromosome Location
Chr. 8: 98189826 - 98295427 on Build GRCh38
UniGene ID
Hs.309489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321635.1 NP_001308564.1 1, 2 392
NM_001321636.1 NP_001308565.1 1, 2 392
NM_024759.2 NP_079035.1 1, 2 392
NR_135745.1 1, 2 392
NR_135746.1 1, 2 392
NR_135747.1 1, 2 392
XM_011517302.2 XP_011515604.1 1, 2 1426
GenBank mRNA AK024017.1 - 1, 2 392
AW082976.1 - 1, 2 182
BC132687.1 - 1, 2 335 40146709
BC144055.1 - 1, 2 335 9052575
DA184376.1 - 1, 2 71

View Full Product Details