Gene Transcripts
- Gene Symbol
- TCTN2
- Entrez Gene ID
- 79867
- Gene Name
- tectonic family member 2
- Gene Aliases
- C12orf38, JBTS24, MKS8, TECT2
- Chromosome Location
- Chr. 12: 123671108 - 123708405 on Build GRCh38
- UniGene ID
- Hs.167165
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143850.2 |
NP_001137322.1
|
13, 14 |
1638 |
|
NM_024809.4 |
NP_079085.2
|
13, 14 |
1641 |
|
XM_005253623.2 |
XP_005253680.1
|
12, 13 |
1506 |
|
XM_006719605.3 |
XP_006719668.1
|
13, 14 |
1641 |
|
XM_017019974.1 |
XP_016875463.1
|
12, 13 |
1503 |
|
XM_017019975.1 |
XP_016875464.1
|
12, 13 |
1516 |
|
GenBank mRNA |
AK023037.1 |
- |
2, 3 |
197 |
|
AK056924.1 |
- |
13, 14 |
1639 |
|
AK292153.1 |
- |
13, 14 |
1638 |
|
BC009112.1 |
- |
13, 14 |
1610 |
3463475 |
DQ278870.1 |
- |
13, 14 |
1639 |
|
View Full Product Details