Gene Transcripts

Gene Symbol
TTC21B
Entrez Gene ID
79809
Gene Name
tetratricopeptide repeat domain 21B
Gene Aliases
ATD4, IFT139, IFT139B, JBTS11, NPHP12, Nbla10696, SRTD4, THM1
Chromosome Location
Chr. 2: 165873362 - 165953838 on Build GRCh38
UniGene ID
Hs.310672
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024753.4 NP_079029.3 28, 29 4014
XM_011511871.2 XP_011510173.1 23, 24 7259
XM_011511872.2 XP_011510174.1 28, 29 3988
XM_017004968.1 XP_016860457.1 23, 24 5505
XM_017004969.1 XP_016860458.1 26, 27 3674
GenBank mRNA AK096451.1 - 12, 13 1641
AK292942.1 - 28, 29 3957
BC035767.2 - 5, 6 780
BC055424.1 - 28, 29 4014 4837489
BC063579.1 - 10, 11 1358
BX647903.1 - 14, 15 1765

View Full Product Details