Gene Transcripts
- Gene Symbol
- POU3F4
- Entrez Gene ID
- 5456
- Gene Name
- POU class 3 homeobox 4
- Gene Aliases
- BRAIN-4, BRN-4, BRN4, DFN3, DFNX2, OCT-9, OTF-9, OTF9
- Chromosome Location
- Chr. X: 83508261 - 83509767 on Build GRCh38
- UniGene ID
- Hs.2229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000307.4 |
NP_000298.3
|
1 |
1480 |
|
GenBank mRNA |
X82324.1 |
- |
1 |
1454 |
|
View Full Product Details