Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
79863
Gene Name
ribosome binding factor A (putative)
Gene Aliases
C18orf22, HsT169
Chromosome Location
Chr. 18: 80034346 - 80050652 on Build GRCh38
UniGene ID
Hs.728931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171967.1 NP_001165438.1 6 831
NM_024805.2 NP_079081.2 7 916
GenBank mRNA AF261073.2 - 7 796
AF439745.2 - 6 711
AK024825.1 - 7 904
BC014195.2 - 7 778 3938406
BC057772.1 - 7 784 30337652

View Full Product Details